Potassium hexacyanoferrate(II) trihydrate 98.5-102.0% ACS

Supplier: Thermo Scientific Chemicals

Synonyms: Potassiumhexacyanoferrate(II) trihydrate, Tetrapotassium hexacyanoferrate trihydrate, Potassium ferrocyanide trihydrate

33358-22 33358-36 33358-A3
CAAA33358-22EA 74.26 CAD
CAAA33358-22 CAAA33358-36 CAAA33358-A3
Potassium hexacyanoferrate(II) trihydrate 98.5-102.0% ACS
Potassium hexacyanoferrate (II) trihydrate

MDL: MFCD00167023 Notes: Begins to lose water at 60°C, anhydrous at 100°C


Purity: 98.5-102.0% CAS No.: 14459-95-1 Molecular Formula: K4Fe(CN)6.3H2O Formula Weight: 422.39 (368.36anhy) Physical Form: Crystalline Density: 1.85 Appearance: Pale yellow Odor: Odorless pH: 9.5 Melting Point: 69-71° Merck Reference: 14,7631 MDL No.: MFCD00167023

Formula: K₄Fe(CN)₆·3H₂O
MW: 422.39 g/mol
Melting Pt: 69…71 °C
Density: 1.85
Storage Temperature: Ambient
MDL Number: MFCD00167023
CAS Number: 14459-95-1
EINECS: 237-722-2
Merck Index: 14,7631

Order Now

Specification Test Results

Insoluble matter 0.005%
Cl 0.01%
SO3 Pass test (limit about 0.01%)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR