D-(-)-Mannitol ≥99.0% ACS
Supplier: Thermo Scientific Chemicals
33342-36
33342-A4
CAAA33342-36EA
124.66
CAD
CAAA33342-36
CAAA33342-A4
D-(-)-Mannitol ≥99.0% ACS
D(-)-Mannitol
MDL: MFCD00064287
Beilstein Registry No.: 1721898
Soluble in water, alcohol, pyridine, aniline. Insoluble in ether
CAS No.: 69-65-8
Molecular Formula: HOCH2(CHOH)4CH2OH
Formula Weight: 182.17
Physical Form: Crystalline
Density: 1.489
Appearance: White
Odor: Odorless
Melting Point: 165-167°
Boiling Point: 290-295°/3.5mm
Solubility: Soluble in water, alcohol, pyridine, aniline. Insoluble in ether
Beilstein Reference: 1721898
Merck Reference: 14,5745
MDL No.: MFCD00064287
Formula:
HOH₂C(CH(OH))₄CH₂OH Boiling Pt: 290…295 °C (3.5 mmHg) Melting Pt: 165…167 °C Density: 1.489 Storage Temperature: Ambient |
MDL Number:
MFCD00064287 CAS Number: 69-65-8 EINECS: 200-711-8 Merck Index: 14,5745 |
|
Specification Test Results
Insoluble matter | 0.01% |
Residue after ignition | 0.01% |
Titratable acid | 0.0008meq/g |
Reducing sugars | Pass test |
Heavy Metals (as Pb) | 5 ppm |
Specific rotation at 25° | +23.3° to +24.3° |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...