Manganese(II) sulfate monohydrate 98.0-101.0% ACS
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
Manganese sulphate monohydrate, Manganese sulfate monohydrate, Manganous sulfate monohydrate, Manganese(II) sulphate hydrate, Manganous sulphate monohydrate, Manganese(II) sulfate hydrate
MDL: MFCD00149159
Notes: Loses all water at 400-450°C
Purity: 98.0-101.0%
CAS No.: 10034-96-5
Molecular Formula: MnSO4.H2O
Formula Weight: 169.02 (151.00anhy)
Physical Form: Powder
Density: 2.95
pH: 3-3.5
Merck Reference: 14,5739
UN No.: 3077
MDL No.: MFCD00149159
Formula:
MnSO₄·1H₂O Melting Pt: 700 °C Density: 2.95 Storage Temperature: Ambient |
MDL Number:
MFCD00149159 CAS Number: 10034-96-5 EINECS: 232-089-9 UN: 3077 ADR: 9,III Merck Index: 14,5739 |
Specification Test Results
Insoluble matter | 0.01% |
Loss on ignition | 10.0-12.0 |
Ca | 0.005% |
Cl | 0.005% |
Heavy Metals (as Pb) | 0.002% |
Fe | 0.002% |
MG | 0.00005 |
Ni | 0.0002 |
K | 0.01%% |
NA | 0.05%% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...