Magnesium sulfate, anhydrous ≥99.5%
Supplier: Thermo Scientific Chemicals
33337-36
33337-A3
33337-A9
CAAA33337-36EA
119.92
CAD
CAAA33337-36
CAAA33337-A3
CAAA33337-A9
Magnesium sulfate, anhydrous ≥99.5%
Magnesium sulphate
Suitable for sensitive molecular biology applications
Purity: 99.5% min
CAS No.: 7487-88-9
Molecular Formula: MgSO4
Formula Weight: 120.37
Physical Form: Powder
Density: 2.66
Appearance: White
Odor: Odorless
Melting Point: 1124° dec.
Merck Reference: 14,5691
MDL No.: MFCD00011110
Suitable for sensitive molecular biology applications
Formula:
MgSO₄ Melting Pt: 1124 °C (dec.) Density: 2.66 Storage Temperature: Ambient |
MDL Number:
MFCD00011110 CAS Number: 7487-88-9 EINECS: 231-298-2 Merck Index: 14,5691 |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...