Magnesium hydroxide carbonate tetrahydrate (∼40 - 43.5% MgO), Reagent Grade

Supplier: Thermo Scientific Chemicals

Synonyms: Magnesium carbonate basic, Magnesium carbonate hydroxide, Magnesium carbonate hydrated basic

33333-22 33333-36
CAAA33333-22EA 46.93 CAD
CAAA33333-22 CAAA33333-36
Magnesium hydroxide carbonate tetrahydrate (∼40 - 43.5% MgO), Reagent Grade
Magnesium hydroxide carbonate tetrahydrate

MDL: MFCD00149783


CAS No.: 39409-82-0 Molecular Formula: 4MgCO3.Mg(OH)2.4H2O approx. Formula Weight: 467.67 (395.58anhy) Physical Form: -325 Mesh Powder Appearance: White Odor: Odorless Merck Reference: 14,5660 MDL No.: MFCD00149783

Formula: 4MgCO₃·Mg(OH)₂·4H₂O
MW: 467.67 g/mol (395.58 anhy)
Melting Pt: > 2500 °C
Density: 2.16 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00149783
CAS Number: 12125-28-9
EINECS: 235-192-7
Merck Index: 14,5660

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR