Lead(II) oxide low silver

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Lead monoxide

33330-36 33330-A3
CAAA33330-36EA 62.25 CAD
CAAA33330-36 CAAA33330-A3
Lead(II) oxide low silver
Lead (II) oxide

As storage batteries, ceramic cements and fluxes, pottery and glazes, in chromium pigments, in oil refining, in paints, and in the assay of precious metal ores


CAS No.: 1317-36-8 Molecular Formula: PbO Formula Weight: 223.19 Physical Form: Powder Density: 9.53 Odor: Odorless Merck Reference: 14,5413 UN No.: 3077 MDL No.: MFCD00011164


As storage batteries, ceramic cements and fluxes, pottery and glazes, in chromium pigments, in oil refining, in paints, and in the assay of precious metal ores

Formula: PbO
MW: 223.2 g/mol
Boiling Pt: 1470 °C (1013 hPa)
Melting Pt: 886 °C
Density: 9.53
Storage Temperature: Ambient
MDL Number: MFCD00011164
CAS Number: 1317-36-8
EINECS: 215-267-0
UN: 3077
ADR: 9,III
Merck Index: 14,5413

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR