Calcium chloride, anhydrous ≥93%, porous
Supplier: Thermo Scientific Chemicals
|
Warning
|
33327-36
33327-A3
CAAA33327-36EA
95.75
CAD
CAAA33327-36
CAAA33327-A3
Calcium chloride, anhydrous ≥93%, porous
Calcium chloride
MDL: MFCD00010903
Purity: 93%
CAS No.: 10043-52-4
Molecular Formula: CaCl2
Formula Weight: 110.99
Physical Form: -4 Mesh
Sensitivity: Hygroscopic
Boiling Point:
Formula:
CaCl₂ Boiling Pt: >1600 °C Melting Pt: 782 °C Density: 2.174 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00010903 CAS Number: 10043-52-4 EINECS: 233-140-8 REACH: 01-2119494219-28 Merck Index: 14,1659 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...