Hydrogen peroxide 29 - 32% w/w aqueous solution Stabilised ACS

Supplier: Thermo Scientific Chemicals
Danger

33323-AD 33323-AP 33323-AY
CAAA33323-ADEA 87.22 CAD
CAAA33323-AD CAAA33323-AP CAAA33323-AY
Hydrogen peroxide 29 - 32% w/w aqueous solution Stabilised ACS
Hydrogen peroxide

For bleaching and deodorizing textiles, wood, hair, etc; rocket propulsion; maturing agent in food; water and sewage treatment; disinfectant; laboratory reagent


CAS No.: 7722-84-1 Molecular Formula: H2O2 Formula Weight: 34.02 Physical Form: Liquid Density: 1.11 Refractive Index: 1.3350 Boiling Point: 107° Solubility: Soluble in water, alcohol and ether. Insoluble in petroleum Beilstein Reference: 3587191 Merck Reference: 14,4798 UN No.: 2014 MDL No.: MFCD00011333


For bleaching and deodorizing textiles, wood, hair, etc; rocket propulsion; maturing agent in food; water and sewage treatment; disinfectant; laboratory reagent

Formula: H₂O₂
MW: 34.01
Boiling Pt: 150.2
Melting Pt: -0.41
Density: 1.11
Storage Temperature: Ambient
MDL Number: MFCD00011333
CAS Number: 7722-84-1
EINECS: 231-765-0
UN: 2014
ADR: 5.1 II
Merck Index: 14,04798

Order Now

Specification Test Results

Color (APHA) 10
Evaporation residue 0.002% max.
Titratable acid 0.0006 meq/g max.
Cl 3 ppm max.
NO2 2 ppm max.
PO3 2 ppm max.
SO3 5 ppm max.
NH 5 ppm max.
Heavy Metals (as Pb) 1 ppm max.
Fe 0.5ppm max.


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR