Iron(III) sulfate hydrate, REacton®, Reagent Grade
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
Ferric Persulphate, Diiron trissulfate hydrate, Diiron trissulphate hydrate, Ferric sulfate hydrate, Ferric sulphate hydrate, Ferric Persulfate
33316-30
33316-A1
33316-A7
CAAA33316-30EA
87.22
CAD
CAAA33316-30
CAAA33316-A1
CAAA33316-A7
Iron(III) sulfate hydrate, REacton®, Reagent Grade
Iron (III) sulphate hydrate
MDL: MFCD00011007
Soluble in water
CAS No.: 15244-10-7
Molecular Formula: Fe2(SO4)3.xH2O
Formula Weight: 399.87(anhy)
Physical Form: Granular/Powder
Density: 2.0-2.1
Sensitivity: Hygroscopic
Appearance: Grayish white
Solubility: Soluble in water
MDL No.: MFCD00011007
Formula:
Fe₂(SO₄)₃·H₂O Density: 2.0…2.1 Storage Temperature: Ambient |
MDL Number:
MFCD00011007 CAS Number: 15244-10-7 EINECS: 233-072-9 Merck Index: 13,04065 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...