Formaldehyde 37% aqueous solution Stabilised ACS
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
Paraformaldehyde aqueous solution, Methanal solution, Methylaldehyde solution, Formol, Formaline solution, Formalin, Methylaldehyde
Stabilization:
10-15% MeOH
MDL: MFCD00003274
Beilstein Registry No.: 1209228
Fieser: 1,397 11,240 12,232 13,136 14,168 15,161 16,171 21,193
CAS No.: 50-00-0
Molecular Formula: CH2O
Formula Weight: 30.03
Flash Point: 60°(140°F)
Physical Form: Liquid
Density: 1.083
Refractive Index: 1.3765
Appearance: Colorless
Odor: Pungent
Melting Point: -15°
Boiling Point: 97°
Beilstein Reference: 1209228
Merck Reference: 14,4235
UN No.: 1198
MDL No.: MFCD00003274
Formula:
CH₂O MW: 30.03 g/mol Boiling Pt: -19.3 Melting Pt: -92 Density: 1.083 Flash Pt: 60 °C (140 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00003274 CAS Number: 50-00-0 EINECS: 200-001-8 UN: 1198 ADR: 3 III Merck Index: 14,4235 |
Specification Test Results
Color (APHA) | 10 |
Residue after ignition | 0.005% max. |
Titratable acid | 0.006meq/g max. |
Cl | 5 ppm max. |
SO3 | 0.002% max. |
Heavy Metals (as Pb) | 5 ppm max. |
Fe | 5 ppm max. |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...