EDTA disodium salt dihydrate 99.0-101.0% ACS
Supplier: Thermo Scientific Chemicals
Synonyms:
Ethylenediaminetetraacetic acid, disodium salt, dihydrate, Ethylenediaminetetraacetic acid disodium salt dihydrate, Kestranal® 2S dihydrate, Disodium dihydrogen ethylenediaminetetraacetate dihydrate, Ethylenedinitrilotetra-acetic acid disodium salt dihydrate, Ethylenediaminetetraacetic acid disodium dihydrate, Titriplex III, Ethylenediaminetetra-acetic acid disodium salt dihydrate, Kestranal® 2S 2 H2O, Sequestrene Na₂, Edetate disodium dihydrate, Edetic acid disodium salt dihydrate
MDL: MFCD00150037
Beilstein Registry No.: 3900609
Fieser: 1,373
Purity: 99.0-101.0%
CAS No.: 6381-92-6
Molecular Formula: (NaO2CCH2)2NCH2CH2N(CH2CO2H)2.2H2O
Formula Weight: 372.24 (336.22anhy)
Physical Form: Powder
Melting Point: 248° dec.
Beilstein Reference: 3900609
MDL No.: MFCD00150037
Formula:
C₁₀H₁₄N₂Na₂O₈·2H₂O MW: 372.24 g/mol Melting Pt: 255 (dec.) Storage Temperature: Ambient |
MDL Number:
MFCD00150037 CAS Number: 6381-92-6 EINECS: 205-358-3 Merck Index: 13,03543 |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...