Cyclohexanone 99+% ACS

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Pimelic Ketone

33309-AP 33309-K2 33309-K7 33309-M1
CAAA33309-APEA 63.83 CAD
CAAA33309-AP CAAA33309-K2 CAAA33309-K7 CAAA33309-M1
Cyclohexanone 99+% ACS
Cyclohexanone

MDL: MFCD00001625 Beilstein Registry No.: 385735 Fieser: 1,178


Purity: 99+% CAS No.: 108-94-1 Molecular Formula: C6H10O Formula Weight: 98.15 Flash Point: 47°(116°F) Physical Form: Liquid Density: 0.9478 Refractive Index: 1.4507 Melting Point: -26° Boiling Point: 154-156° Beilstein Reference: 385735 Merck Reference: 14,2726 UN No.: 1915 MDL No.: MFCD00001625

Formula: C₆H₁₀(=O)
MW: 98.14 g/mol
Boiling Pt: 154…156 °C
Melting Pt: –26 °C
Density: 0.9478
Flash Pt: 47 °C (116 °F)
Storage Temperature: Ambient
MDL Number: MFCD00001625
CAS Number: 108-94-1
EINECS: 203-631-1
UN: 1915
ADR: 3,III
REACH: 01-2119453616-35
Merck Index: 14,2726
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=b8701dac7edb70a452483cd4f1bd716ca29ba9aa&vid=0360daffe7606174162954ba58a6ba2b78e28dfb&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Color (APHA) 10
Evaporation residue 0.0005

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR