Copper(II) sulfate, anhydrous, REacton®, Reagent Grade

Supplier: Thermo Scientific Chemicals
Warning

33308-22 33308-36 33308-A3
CAAA33308-22EA 88.8 CAD
CAAA33308-22 CAAA33308-36 CAAA33308-A3
Copper(II) sulfate, anhydrous, REacton®, Reagent Grade
Copper (II) sulphate

MDL: MFCD00010981 Soluble in water. Insoluble in alcohol


CAS No.: 7758-98-7 Molecular Formula: CuO4S Formula Weight: 159.60 Physical Form: Powder Density: 3.6 Sensitivity: Hygroscopic Appearance: Pale grey Odor: Odorless Melting Point: 560° dec. Solubility: Soluble in water. Insoluble in alcohol Merck Reference: 14,2653 UN No.: 3288 MDL No.: MFCD00010981

Formula: CuSO₄
Melting Pt: >560 °C (dec.)
Density: 3.6
Storage Temperature: Ambient
MDL Number: MFCD00010981
CAS Number: 7758-98-7
EINECS: 231-847-6
UN: 3288
ADR: 6.1,III
Merck Index: 14,2653

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR