Charcoal activated, powder (total ash max 4%)
Supplier: Thermo Scientific Chemicals
|
Danger
|
33302-22
33302-36
CAAA33302-22EA
57.83
CAD
CAAA33302-22
CAAA33302-36
Charcoal activated, powder (total ash max 4%)
Charcoal activated
MDL: MFCD00133992
CAS No.: 7440-44-0
Appearance: Black
UN No.: 1362
MDL No.: MFCD00133992
Formula:
C Melting Pt: 3550 °C Density: 2 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00133992 CAS Number: 7440-44-0 EINECS: 231-153-3 UN: 1362 ADR: 4.2,III REACH: 01-2119488894-16 Merck Index: 13,01818 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...