Calcium sulfate dihydrate 99%
Supplier: Thermo Scientific Chemicals
33301-A1
33301-A7
CAAA33301-A1EA
133.67
CAD
CAAA33301-A1
CAAA33301-A7
Calcium sulfate dihydrate 99%
Calcium sulphate dihydrate
MDL: MFCD00149625
Purity: 99%
CAS No.: 10101-41-4
Molecular Formula: CaSO4.2H2O
Formula Weight: 172.17 (136.14anhy)
Physical Form: Powder
Density: 2.32
Melting Point: 128° -1.5H2O
Boiling Point: 163° -2H2O
Merck Reference: 14,1706
MDL No.: MFCD00149625
Formula:
CaSO₄·2H₂O Boiling Pt: 163 °C (-2H₂O) Melting Pt: 128 °C (-1.5H₂O) Density: 2.32 Storage Temperature: Ambient |
MDL Number:
MFCD00149625 CAS Number: 10101-41-4 EINECS: 231-900-3 Merck Index: 14,1706 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...