Arsenic(III) oxide 99.95-100.05% ACS primary standard
Supplier: Thermo Scientific Chemicals
|
Danger
|
33289-14
33289-22
CAAA33289-14EA
162.74
CAD
CAAA33289-14
CAAA33289-22
Arsenic(III) oxide 99.95-100.05% ACS primary standard
Arsenic (III) oxide
MDL: MFCD00003433
Purity: 99.95-100.05
CAS No.: 1327-53-3
Molecular Formula: As2O3
Formula Weight: 197.84
Physical Form: Powder
Density: 3.74
Melting Point: 312.3°
Boiling Point: 465° subl.
Merck Reference: 14,804
UN No.: 1561
MDL No.: MFCD00003433
Formula:
As₂O₃ Boiling Pt: 465 °C (subl.) Melting Pt: 312.3 °C Density: 3.74 Storage Temperature: Ambient |
MDL Number:
MFCD00003433 CAS Number: 1327-53-3 EINECS: 215-481-4 UN: 1561 ADR: 6.1,II Merck Index: 14,804 |
Specification Test Results
Residue after ignition | 0.02% |
Insoluble in dilute hydrochloric acid | 0.01% |
Cl | 0.005% |
S | Pass test (Limit about 0.001%) |
Sb | 0.05% |
PB | 0.002% |
Fe | 5 ppm max. |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...