Ammonium sulfide 20 - 24% in aqueous solution

Supplier: Thermo Scientific Chemicals
Danger

33286-AE 33286-AP
CAAA33286-AEEA 128.3 CAD
CAAA33286-AE CAAA33286-AP
Ammonium sulfide 20 - 24% in aqueous solution
Ammonium sulphide

MDL: MFCD00010892


CAS No.: 12135-76-1 Molecular Formula: H8N2S Formula Weight: 68.14 Physical Form: Liquid Density: 0.997 Odor: Like rotten eggs UN No.: 2683 MDL No.: MFCD00010892

Formula: H₈N₂S
Density: 0.997
Storage Temperature: Ambient
MDL Number: MFCD00010892
CAS Number: 12135-76-1
EINECS: 235-223-4
UN: 2683
ADR: 8,II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR