Ammonia solution 28.0 - 30.0% ACS
Supplier: Thermo Scientific Chemicals
|
Danger
|
33285-30
33285-A1
33285-D9
CAAA33285-30EA
49.3
CAD
CAAA33285-30
CAAA33285-A1
CAAA33285-D9
Ammonia solution 28.0 - 30.0% ACS
Ammonia solution
MDL: MFCD00066650
CAS No.: 1336-21-6
Molecular Formula: NH4OH
Formula Weight: 35.05
Physical Form: Liquid
Density: 0.90
Appearance: Colorless
Odor: Ammonia-like
pH: 12
Merck Reference: 14,494
UN No.: 2672
MDL No.: MFCD00066650
Formula:
NH₃ Boiling Pt: 32…37.7 °C (1013 hPa) Melting Pt: –60…–57.5 °C Density: 0.90 Storage Temperature: Ambient |
MDL Number:
MFCD00066650 CAS Number: 1336-21-6 EINECS: 215-647-6 UN: 2672 ADR: 8,III Merck Index: 14,494 |
Specification Test Results
Residue after ignition | 0.002% |
CO2 | 0.002% |
Cl | 0.5 ppm |
PO3 | 2 ppm |
NO3 | 2 ppm |
Sulfate (SO3) | 2 ppm |
Fe | 0.2 ppm |
Substances reducing permanganate | Pass test |
Heavy Metals (as Pb) | 0.5 ppm |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...