Dithizone ≥85% ACS

Supplier: Thermo Scientific Chemicals
Warning

33283-04 33283-09 33283-18
CAAA33283-04EA 43.92 CAD
CAAA33283-04 CAAA33283-09 CAAA33283-18
Dithizone ≥85% ACS
Dithizone

MDL: MFCD00003025 Beilstein Registry No.: 748838


CAS No.: 60-10-6 Molecular Formula: C13H12N4S Formula Weight: 256.33 Physical Form: Crystalline Odor: Odorless Melting Point: 168° dec. Beilstein Reference: 748838 Merck Reference: 14,3377 MDL No.: MFCD00003025

Formula: C₆H₅NHNHCSN=NC₆H₅
Boiling Pt: 406 °C (1013 hPa)
Melting Pt: 168 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00003025
CAS Number: 60-10-6
EINECS: 200-454-1
Merck Index: 14,3377

Order Now

Specification Test Results

Residue after ignition 0.3%
Heavy Metals (as Pb) 0.002% (as Pb)
Ratio of absorbance 1.55 Ratio


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR