tert-Butanol 99+% ACS
Supplier: Thermo Scientific Chemicals
|
Danger
|
33278-AK
33278-K2
33278-K7
33278-M1
CAAA33278-AKEA
55.62
CAD
CAAA33278-AK
CAAA33278-K2
CAAA33278-K7
CAAA33278-M1
tert-Butanol 99+% ACS
tert-Butanol
MDL: MFCD00004464
Beilstein Registry No.: 906698
Purity: 99+%
CAS No.: 75-65-0
Molecular Formula: C4H10O
Formula Weight: 74.12
Flash Point: 11°(52°F)
Physical Form: Liquid
Density: 0.786
Refractive Index: 1.3870
Appearance: Colorless
Melting Point: 25°
Boiling Point: 83°
Beilstein Reference: 906698
Merck Reference: 14,1542
UN No.: 1120
MDL No.: MFCD00004464
Formula:
(CH₃)₃COH Boiling Pt: 83 °C Melting Pt: 25 °C Density: 0.786 Flash Pt: 11 °C (52 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00004464 CAS Number: 75-65-0 EINECS: 200-889-7 UN: 1120 ADR: 3,II Merck Index: 14,1542 |
|
Specification Test Results
Color (APHA) | 20 |
Residue after evaporation | 0.3% |
Titratable acid | 0.001meq/g |
Carbonyl compounds (as formaldehyde) | 0.01% |
H2O | 0.1% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...