Sulfuric acid 95.0-98.0% ACS

Supplier: Thermo Scientific Chemicals
Danger

33273-A1 33273-D6
CAAA33273-A1EA 104.91 CAD
CAAA33273-A1 CAAA33273-D6
Sulfuric acid 95.0-98.0% ACS
Sulfuric acid

MDL: MFCD00064589


Purity: 95.0-98.0% CAS No.: 7664-93-9 Molecular Formula: H2SO4 Formula Weight: 98.08 Physical Form: Liquid Density: 1.84 Appearance: Colorless to pale yellow Odor: Acrid Merck Reference: 14,8974 UN No.: 1830 MDL No.: MFCD00064589

Formula: H₂SO₄
MW: 98.08
Boiling Pt: 330
Melting Pt: 10.38
Density: 1.84
Storage Temperature: Ambient
MDL Number: MFCD00064589
CAS Number: 7664-93-9
EINECS: 231-639-5
UN: 1830
ADR: 8 II
Merck Index: 14,8974

Order Now

Specification Test Results

Color (APHA) 10
Residue after ignition 5 ppm
Cl 0.2 ppm
NO3 0.5 ppm
NH3 2 ppm
Substances reducing permanganate (as SO2) 2 ppm
As 0.01 ppm
Heavy metals (as Pb) 1 ppm
Fe 0.2 ppm
Hg 5ppb


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR