Succinic acid ≥99.0% ACS

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: 1,4-Butanedioic acid

33272-36 33272-A3
CAAA33272-36EA 103.96 CAD
CAAA33272-36 CAAA33272-A3
Succinic acid ≥99.0% ACS
Succinic acid

MDL: MFCD00002789 Beilstein Registry No.: 1754069 Soluble in water (increasing with temperature). Soluble in alcohol, methanol


Purity: 99.0% min CAS No.: 110-15-6 Molecular Formula: HO2CCH2CH2CO2H Formula Weight: 118.09 Flash Point: 206°(402°F) Physical Form: Crystalline Density: 1.56 Melting Point: 186-188° Boiling Point: 235° Solubility: Soluble in water (increasing with temperature). Soluble in alcohol, methanol Beilstein Reference: 1754069 Merck Reference: 14,8869 MDL No.: MFCD00002789

Formula: HOOCCH₂CH₂COOH
MW: 118.09 g/mol
Boiling Pt: 235 °C
Melting Pt: 186…188 °C
Density: 1.56
Flash Pt: 206 °C (402 °F)
Storage Temperature: Ambient
MDL Number: MFCD00002789
CAS Number: 110-15-6
EINECS: 203-740-4
UN: 3261
ADR: 8,III
Merck Index: 14,8869
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=dfd381e6e0e4cbde676a92c9e8d6aacfac635ade&vid=0b14e1b7f82b03468fc82d224a0ee894952eb200&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Insoluble matter 0.01%
Residue after ignition 0.02%
Cl 0.001%
PO3 0.00001
SO3 0.003%
Nitrogen compounds (as N) 0.001%
Heavy Metals (as Pb) 5 ppm max.
Fe 5 ppm
Melting point 185.0°-191.0°

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR