Orthophosphoric acid ≥85% (w/w) in aqueous solution ACS
Supplier: Thermo Scientific Chemicals
|
Danger
|
33266-22
33266-A1
33266-D6
CAAA33266-22EA
56.88
CAD
CAAA33266-22
CAAA33266-A1
CAAA33266-D6
Orthophosphoric acid ≥85% (w/w) in aqueous solution ACS
Orthophosphoric acid
MDL: MFCD00011340
Fieser: 1,860 4,387 7,255 10,317 15,266
CAS No.: 7664-38-2
Molecular Formula: H3O4P
Formula Weight: 98.00
Physical Form: Viscous liquid
Density: 1.70
Refractive Index: 1.4310
Melting Point: 19-21°
Merck Reference: 14,7344
UN No.: 1805
MDL No.: MFCD00011340
Formula:
H₃PO₄ Melting Pt: 19…21 °C Density: 1.70 Storage Temperature: Ambient |
MDL Number:
MFCD00011340 CAS Number: 7664-38-2 EINECS: 231-633-2 UN: 1805 ADR: 8,III Merck Index: 14,7344 |
Specification Test Results
Color (APHA) | 10 |
Insoluble matter | 0.001% |
Cl | 3 ppm |
NO3 | 5 ppm |
SO3 | 0.003% |
Volatile acids (as CH3COOH) | 0.001% |
Sb | 0.002% |
Ca | 0.002% |
MG | 0.002% |
As | 1ppm |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...