Oxalic acid dihydrate 99.5-102.5% ACS

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Ethanedioic acid dihydrate

33262-18 33262-30 33262-A1 33262-A7
CAAA33262-18EA 60.36 CAD
CAAA33262-18 CAAA33262-30 CAAA33262-A1 CAAA33262-A7
Oxalic acid dihydrate 99.5-102.5% ACS
Oxalic acid dihydrate

As an analytical reagent, in dye and paint industries, in polymers, in metal treatment, in cleaning and textile finishing, and as a reagent for dehydration and condensation


Purity: 99.5-102.5% CAS No.: 6153-56-6 Molecular Formula: C2H2O4.2H2O Formula Weight: 126.07 (90.04anhy) Physical Form: Crystalline Density: 1.65 Appearance: White Odor: Odorless pH: 1.3 Melting Point: 101-105° Solubility: Very soluble in water. Moderately soluble in ethanol. Sparingly soluble in ether Beilstein Reference: 3679436 Merck Reference: 14,6911 UN No.: 3261 MDL No.: MFCD00149102


As an analytical reagent, in dye and paint industries, in polymers, in metal treatment, in cleaning and textile finishing, and as a reagent for dehydration and condensation

Formula: HO₂CCO₂H·2H₂O
MW: 126.07 g/mol
Boiling Pt: 149…160 °C (1013 hPa)
Melting Pt: 101…105 °C
Density: 1.65
Storage Temperature: Ambient
MDL Number: MFCD00149102
CAS Number: 6153-56-6
EINECS: 205-634-3
UN: 3261
ADR: 8,III
REACH: 01-2119534576-33
Merck Index: 14,6911
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=dcc1447f5f23a589dc85e10dd090c86e33b6541b&vid=2f0e34543b2f625715b002905fc0dd45fcd75561&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Insoluble matter 0.005
Residue after ignition 0.01%
Cl 0.002%
SO3 0.005%
Ca 0.001%
Nitrogen compounds (as N) 0.001%
Heavy Metals (as Pb) 5 ppm
Fe 2 ppm
Substances darkened by hot sulfuric acid Pass test

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR