Acetic acid glacial_AcOH 99
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
Vinegar acid, Methanecarboxylic acid, Ethanoic acid
33252-AK
33252-K2
33252-K7
33252-M4
33252-M6
CAAA33252-AKEA
70.31
CAD
CAAA33252-AK
CAAA33252-K2
CAAA33252-K7
CAAA33252-M4
CAAA33252-M6
Acetic acid glacial_AcOH 99
Acetic acid
MDL: MFCD00036152
Fieser: 2,5 5,3 7,1 8,1 19,1 21,1
Purity: 99+%
CAS No.: 64-19-7
Molecular Formula: CH3CO2H
Formula Weight: 60.05
Flash Point: 40°(104°F)
Physical Form: Liquid
Density: 1.049
Refractive Index: 1.3721
Odor: Pungent
pH: 2,5
Melting Point: 16.6°
Boiling Point: 118.1°
Merck Reference: 14,55
UN No.: 2789
MDL No.: MFCD00036152
Formula:
H₃CCOOH MW: 60.05 g/mol Boiling Pt: 118 Melting Pt: 17 Density: 1.049 Flash Pt: 40 °C (104 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00036152 CAS Number: 64-19-7 EINECS: 200-580-7 UN: 2789 ADR: 8 II Merck Index: 14,55 |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...