Cadmium oxide ≥98.9%
Supplier: Thermo Scientific Chemicals
|
Danger
|
33235-30
33235-A1
CAAA33235-30EA
75.37
CAD
CAAA33235-30
CAAA33235-A1
Cadmium oxide ≥98.9%
Cadmium oxide
MDL: MFCD00010921
Practically insoluble in water. Soluble in dilute acids, slowly soluble in ammonium salts.
Purity: 98.9%
CAS No.: 1306-19-0
Molecular Formula: CdO
Formula Weight: 128.40
Physical Form: Powder
Density: 8.15
Appearance: Red-brown
Odor: Odorless
Melting Point: 1430°
Solubility: Practically insoluble in water. Soluble in dilute acids, slowly soluble in ammonium salts
Merck Reference: 14,1623
UN No.: 2570
MDL No.: MFCD00010921
Formula:
CdO Boiling Pt: 1559 °C (1013 hPa) Melting Pt: 1430 °C Density: 8.15 Storage Temperature: Ambient |
MDL Number:
MFCD00010921 CAS Number: 1306-19-0 EINECS: 215-146-2 UN: 2570 ADR: 6.1,III Merck Index: 14,1623 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...