Potassium citrate monohydrate 99+%

Supplier: Thermo Scientific Chemicals

33222-36 33222-A3
CAAA33222-36EA 70.31 CAD
CAAA33222-36 CAAA33222-A3
Potassium citrate monohydrate 99+%
Potassium citrate monohydrate

MDL: MFCD00150442 Soluble in water and glycerol. Practically insoluble in alcohol


Purity: 99+% CAS No.: 6100-05-6 Molecular Formula: K3C6H5O7.H2O Formula Weight: 324.42 (306.41anhy) Physical Form: Granular Density: 1.98 Melting Point: 230° dec. Solubility: Soluble in water and glycerol. Practically insoluble in alcohol Merck Reference: 14,7623 MDL No.: MFCD00150442

Formula: HOC(COOK)(CH₂COOK)₂·H₂O
Melting Pt: 230 °C (dec.)
Density: 1.98
Storage Temperature: Ambient
MDL Number: MFCD00150442
CAS Number: 6100-05-6
EINECS: 212-755-5
Merck Index: 14,7623
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=dc3e6bae942870a760ea6cb418203db7458ff3dc&vid=2c2580a7bfc83e60c4dfca9f8af4ac3b3db56b27&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR