Diammonium hydrogen citrate 99+%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
Citric acid diammonium salt, Diammonium hydrogen 2-hydroxypropane-1,2,3-tricarboxylate, Ammonium citrate dibasic
33221-22
33221-36
33221-A3
CAAA33221-22EA
71.89
CAD
CAAA33221-22
CAAA33221-36
CAAA33221-A3
Diammonium hydrogen citrate 99+%
di-Ammonium hydrogen citrate
MDL: MFCD00013068
Purity: 99+%
CAS No.: 3012-65-5
Molecular Formula: C6H14N2O7
Formula Weight: 226.19
Physical Form: Granular
Density: 1.48
Appearance: White
Odor: Odorless
Merck Reference: 14,512
MDL No.: MFCD00013068
Formula:
HOC(CO₂H)(CH₂CO₂NH₄)₂ Density: 1.48 Storage Temperature: Ambient |
MDL Number:
MFCD00013068 CAS Number: 3012-65-5 EINECS: 221-146-3 Merck Index: 14,512 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...