Bromine ≥99.5%, liquid ACS

Supplier: Thermo Scientific Chemicals

33220-61 33220-22 33220-09 33220-61 33220-22 33220-09 33220-36 33220-36
CAAA33220-22EA 118.34 CAD
AA33220-61 CAAA33220-22 AA33220-22 CAAA33220-36 AA33220-36 CAAA33220-09 CAAA33220-61 AA33220-09
Bromine ≥99.5%, liquid ACS
Bromine
Formula: Br₂
MW: 159.81 g/mol
Boiling Pt: 58.7 °C (1013 hPa)
Melting Pt: –7.2 °C
Density: 3.12 g/cm³ (20 °C)
MDL Number: MFCD00010896
CAS Number: 7726-95-6
UN: 1744
ADR: 8,I
Merck Index: 13,01378

Order Now

Specification Test Results

Evaporation residue 0.005% Max.
Cl 0.05% Max.
I 0.001% Max.
Sulfur compounds (as S) 0.001% Max.
Heavy Metals (as Pb) 2 ppm Max.
Ni 5 ppm Max.

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR