Bismuth sodium trioxide ≥80% ACS

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Sodium bismuthate, Sodium bismuth oxide

33219-14 33219-22
CAAA33219-14EA 132.25 CAD
CAAA33219-14 CAAA33219-22
Bismuth sodium trioxide ≥80% ACS
Bismuth sodium trioxide

Manganese testing in iron, steel, ores


Purity: 80% min CAS No.: 12232-99-4 Molecular Formula: NaBiO3 Formula Weight: 279.97 Physical Form: Powder Sensitivity: Hygroscopic Appearance: Yellow to brown Odor: Odorless Solubility: Insoluble in cold water. Decomposes in hot water forming Bi2O3, NaOH, and oxygen Merck Reference: 14,8585 MDL No.: MFCD00003474


Manganese testing in iron, steel, ores

Formula: BiNaO₃
MW: 279.97 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00003474
CAS Number: 12232-99-4
EINECS: 235-455-6
Merck Index: 14,8585

Order Now

Specification Test Results

Oxidizing efficiency 99.6 %
minimum Cl 0.002%
Mn 5 ppm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR