Wood's alloy (lipowitz's alloy) (Bi:Pb:Sn:Cd 50:25:12.5;12.5 wt%), ingot
Supplier: Thermo Scientific Chemicals
|
Danger
|
33218-22
33218-36
33218-A3
CAAA33218-22EA
77.74
CAD
CAAA33218-22
CAAA33218-36
CAAA33218-A3
Wood's alloy (lipowitz's alloy) (Bi:Pb:Sn:Cd 50:25:12.5;12.5 wt%), ingot
Wood's alloy (Lipowitz's alloy)
MDL: MFCD00198175
CAS No.: 8049-22-7
Molecular Formula: Bi:Pb:Sn:Cd; 50:25:12.5:12.5 wt%
Physical Form: Ingot, fusible alloy
Appearance: Grey
Odor: Odorless
Melting Point: 71°
UN No.: 3077
MDL No.: MFCD00198175
Melting Pt:
71 °C Density: 9.7 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00198175 CAS Number: 76093-98-6 UN: 3077 ADR: 9,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...