Bismuth(III) carbonate basic ≥98.5%

Supplier: Thermo Scientific Chemicals

Synonyms: Bismuth carbonate oxide, Dibismuth carbonate dioxide, Bismuth subcarbonate

33216-22 33216-36
CAAA33216-22EA 111.39 CAD
CAAA33216-22 CAAA33216-36
Bismuth(III) carbonate basic ≥98.5%
Bismuth (III) carbonate basic

Opacifier in X-ray diagnosis, enamel fluxes, ceramic glazes


Purity: 98.5% min CAS No.: 5892-10-4 Molecular Formula: (BiO)2CO3 (approximate) Formula Weight: 509.97 Physical Form: Powder Density: 6.86 Solubility: Practically insoluble in water, alcohol. Soluble in nitric acid, hydrochloric acid, concentrated acetic acid Merck Reference: 14,1282 MDL No.: MFCD00010877


Opacifier in X-ray diagnosis, enamel fluxes, ceramic glazes

Formula: (BiO)₂CO₃
MW: 509.97 g/mol
Density: 6.86
Storage Temperature: Ambient
MDL Number: MFCD00010877
CAS Number: 5892-10-4
EINECS: 227-567-9
Merck Index: 14,1282

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR