Phenol 99+% stabilised ACS
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
Phenylalcohol, Carbolic acid, Hydroxybenzene
33213-22
33213-36
33213-A3
CAAA33213-22EA
80.3
CAD
CAAA33213-22
CAAA33213-36
CAAA33213-A3
Phenol 99+% stabilised ACS
Phenol
MDL: MFCD00002143
Beilstein Registry No.: 969616
Fieser: 1828
Purity: 99+%
CAS No.: 108-95-2
Molecular Formula: C6H5OH
Formula Weight: 94.11
Flash Point: 79°(175°F)
Physical Form: Crystalline
Density: 1.071
Sensitivity: Air Sensitive
Odor: Sweet
Melting Point: 40-43°
Boiling Point: 181°
Beilstein Reference: 969616
Merck Reference: 14,7241
UN No.: 1671
MDL No.: MFCD00002143
Formula:
C₆H₅OH Boiling Pt: 181 °C Melting Pt: 40…43 °C Density: 1.071 Flash Pt: 79 °C (175 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00002143 CAS Number: 108-95-2 EINECS: 203-632-7 UN: 1671 ADR: 6.1,II REACH: 01-2119471329-32 Merck Index: 14,7241 |
Specification Test Results
Clarity of solution | Pass test |
Evaporation residue | 0.0005 |
H2O | 0.005 |
Freezing point | 40.5° |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...