Aluminium oxide ≥99.7%, fused, insulating powder ∼ 60 µm

Supplier: Thermo Scientific Chemicals

33200-A1 33200-A7
CAAA33200-A1EA 135.41 CAD
CAAA33200-A1 CAAA33200-A7
Aluminium oxide ≥99.7%, fused, insulating powder ∼ 60 µm
Aluminium oxide

Used in dry filling for thermal insulation of electrical furnaces.


CAS No.: 1344-28-1 Molecular Formula: Al2O3 Physical Form: ≈60 micron Odor: Odorless MDL No.: MFCD00003424


Used in dry filling for thermal insulation of electrical furnaces.

Formula: Al₂O₃
Boiling Pt: 2980 °C (1013 hPa)
Melting Pt: 2045 °C
Density: 3.94 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00003424
CAS Number: 1344-28-1
EINECS: 215-691-6
REACH: 01-2119529248-35
Merck Index: 13,00355

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR