4-Methyl-1-pentene 98+%
Supplier: Thermo Scientific Chemicals
|
Danger
|
33029-09
33029-18
33029-30
CAAA33029-09EA
65.1
CAD
CAAA33029-09
CAAA33029-18
CAAA33029-30
4-Methyl-1-pentene 98+%
4-Methyl-1-pentene
MDL: MFCD00008949
Beilstein Registry No.: 1731096
Purity: 98+%
CAS No.: 691-37-2
Molecular Formula: C6H12
Formula Weight: 84.16
Flash Point: -32°(-25°F)
Physical Form: Liquid
Density: 0.664
Refractive Index: 1.3820
Appearance: Colorless
Odor: Petrol-like
Melting Point: -153°
Boiling Point: 53-54°
Beilstein Reference: 1731096
UN No.: 3295
MDL No.: MFCD00008949
Formula:
C₆H₁₂ Boiling Pt: 53…54 °C Melting Pt: –153 °C Density: 0.664 Flash Pt: –32 °C (–25 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00008949 CAS Number: 691-37-2 EINECS: 211-720-1 UN: 3295 ADR: 3,II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...