Cadmium(II) bromide, anhydrous 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Cadmium bromide is used in process engraving, lithography and photography.
Purity: 98%
CAS No.: 7789-42-6
Molecular Formula: CdBr2
Formula Weight: 272.22
Physical Form: Powder
Density: 5.192
Appearance: White to pale yellow
Odor: Odorless
Melting Point: 567°
Boiling Point: 863°
Solubility: Freely soluble in water, alcohol. Slightly soluble in acetone and ether.
Merck Reference: 14,1615
UN No.: 2570
MDL No.: MFCD00015996
Process engraving, lithography and photography
Freely soluble in water, alcohol. Slightly soluble in acetone and ether.
Formula:
Br₂Cd Boiling Pt: 863 °C Melting Pt: 567 °C Density: 5.192 Storage Temperature: Ambient |
MDL Number:
MFCD00015996 CAS Number: 7789-42-6 EINECS: 232-165-1 UN: 2570 ADR: 6.1,III Merck Index: 14,1615 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...