Cadmium(II) bromide tetrahydrate, Reagent Grade

Supplier: Thermo Scientific Chemicals
Warning

32958-14 32958-22
CAAA32958-14EA 73.63 CAD
CAAA32958-14 CAAA32958-22
Cadmium(II) bromide tetrahydrate, Reagent Grade
Cadmium (II) bromide tetrahydrate

MDL: MFCD00150204


CAS No.: 13464-92-1 Molecular Formula: CdBr2.4H2O Formula Weight: 344.28 (272.22anhy) Physical Form: Powder Sensitivity: Hygroscopic MDL No.: MFCD00150204

Formula: Br₂Cd·4H₂O
MW: 344.28
Melting Pt: 575
Density: 5.214 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00150204
CAS Number: 13464-92-1
EINECS: 232-165-1
UN: 2570
ADR: 6.1 III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR