Manganese(II) iodide, anhydrous 98%

Supplier: Thermo Scientific Chemicals
Warning

32808-09 32808-18
CAAA32808-09EA 159.58 CAD
CAAA32808-09 CAAA32808-18
Manganese(II) iodide, anhydrous 98%
Manganese (II) iodide

MDL: MFCD00016225 Soluble in water with gradual decomposition. Soluble in alcohol.


Purity: 98% CAS No.: 7790-33-2 Molecular Formula: I2Mn Formula Weight: 308.75 Physical Form: Powder Density: 5.0 Appearance: White to pink to red Odor: Odorless Melting Point: 638° Solubility: Soluble in water with gradual decomposition. Soluble in alcohol Merck Reference: 14,5732 MDL No.: MFCD00016225

Formula: I₂Mn
Melting Pt: 638 °C
Density: 5.0
Storage Temperature: Ambient
MDL Number: MFCD00016225
CAS Number: 7790-33-2
EINECS: 232-201-6
Merck Index: 14,5732

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR