Manganese(II) bromide, anhydrous 99% H₂O <0.3%, powder -100 Mesh

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Manganous bromide, Manganese(ii)bromide, Manganese dibromide, Manganese(II) bromide hydrate

32736-09 32736-18
CAAA32736-09EA 115.34 CAD
CAAA32736-09 CAAA32736-18
Manganese(II) bromide, anhydrous 99% H₂O <0.3%, powder -100 Mesh
Manganese (II) bromide

It can be used in place of palladium in the Stille reaction, which couples two carbon atoms using an organotin compound.


Hygroscopic. Incompatible with oxidizing and reducing agents. Store at room temperature. Handle under dry protective gas.

Formula: Br₂Mn
MW: 214.75 g/mol
Melting Pt: 698
Density: 4.38
Storage Temperature: Ambient
MDL Number: MFCD00011113
CAS Number: 13446-03-2
EINECS: 236-591-9
Merck Index: 14,5725

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR