Copper(II) hydroxide 94% stabilised, tech.

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Cupric hydroxide, Cuprichydroxide, Copper dihydroxide

32733-36 32733-A3
CAAA32733-36EA 115.5 CAD
CAAA32733-36 CAAA32733-A3
Copper(II) hydroxide 94% stabilised, tech.
Copper (II) hydroxide

Used as a raw material in chemical processing and catalyst manufacturing.


Purity: 94% CAS No.: 20427-59-2 Molecular Formula: H2CuO2 Formula Weight: 97.55 Physical Form: Powder Density: 3.4 Appearance: Blue Odor: Odorless Solubility: Slightly soluble in water Merck Reference: 14,2642 MDL No.: MFCD00010968


Used as a raw material in chemical processing and catalyst manufacturing.

Formula: H₂CuO₂
MW: 97.56
Density: 3.4
Storage Temperature: Ambient
MDL Number: MFCD00010968
CAS Number: 20427-59-2
EINECS: 243-815-9
UN: 3288
ADR: 6.1 II
Merck Index: 14,2642

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR