Hydrazine dihydrochloride 98+%
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
Hydrazine Dihydrochloride, Hydrazinium dichloride
As chlorine scavenger in HCl gas streams
Purity: 98+%
CAS No.: 5341-61-7
Molecular Formula: H2NNH2.2HCl
Formula Weight: 104.97
Physical Form: Lump
Density: 1.42
Appearance: White
Odor: Odorless
Melting Point: 198°
Boiling Point: 200° dec.
Solubility: Soluble in water. Slightly soluble in alcohol
Merck Reference: 14,4770
UN No.: 3288
MDL No.: MFCD00064543
As chlorine scavenger in HCl gas streams
Formula:
H₆Cl₂N₂ MW: 104.97 g/mol Boiling Pt: 200 °C (dec.) Melting Pt: 189 Density: 1.42 Storage Temperature: Ambient |
MDL Number:
MFCD00064543 CAS Number: 5341-61-7 EINECS: 226-283-2 UN: 3288 ADR: 6.1 III Merck Index: 14,4770 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...