Bromophenol blue ACS
Supplier: Thermo Scientific Chemicals
32641-03
32641-06
32641-14
CAAA32641-03EA
33.34
CAD
CAAA32641-03
CAAA32641-06
CAAA32641-14
Bromophenol blue ACS
Bromophenol blue
MDL: MFCD00005875
Beilstein Registry No.: 61698
Freely soluble in NaOH
CAS No.: 115-39-9
Molecular Formula: C19H10Br4O5S
Formula Weight: 669.98
Physical Form: Crystalline
Odor: Odorless
Melting Point: 270° dec.
Beilstein Reference: 61698
Merck Reference: 14,1444
MDL No.: MFCD00005875
Formula:
C₁₉H₁₀Br₄O₅S Melting Pt: 270 °C (dec.) Density: 1 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00005875 CAS Number: 115-39-9 EINECS: 204-086-2 Merck Index: 14,1444 |
Specification Test Results
Clarity of solution | Pass test |
Visual transition interval | pH 3.0 (yellow) to pH 4.6 (blue) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...