Bromophenol blue sodium salt ACS

Supplier: Thermo Scientific Chemicals

Synonyms: Bromophenol blue sodium, Sodium α-(3,5-dibromo-4-oxo-2,5-cyclohexadienylidene)-α-(3,5-dibromo-4-hydroxyphenyl)toluenesulphonate, Sodium 2,6-dibromo-4-(3-(3,5-dibromo-4-hydroxyphenyl)-1,1-dioxido-3H-benzo[c][1,2]oxathiol-3-yl)phenolate, Bromophenol blue water soluble, 4,4'-(3H-2,1-Benzoxathiol-3-ylidene)bis[2,6-dibromophenol] S,S-dioxide sodium salt, Sodium α-(3,5-dibromo-4-oxo-2,5-cyclohexadienylidene)-α-(3,5-dibromo-4-hydroxyphenyl)toluenesulfonate

32639-06 32639-14
CAAA32639-06EA 51.82 CAD
CAAA32639-06 CAAA32639-14
Bromophenol blue sodium salt ACS
Bromophenol blue sodium salt

MDL: MFCD00013793


CAS No.: 34725-61-6 Molecular Formula: C19H9Br4NaO5S Formula Weight: 691.96 Physical Form: Crystalline Melting Point: 300° Merck Reference: 14,1444 MDL No.: MFCD00013793

Formula: C₁₉H₉Br₄NaO₅S
MW: 691.95 g/mol
Melting Pt: >300 °C
Storage Temperature: Ambient
MDL Number: MFCD00013793
CAS Number: 34725-61-6
EINECS: 252-170-2
Merck Index: 14,1444

Order Now

Specification Test Results

Clarity of solution Pass test
Visual transition interval pH 3.0 (yellow) to pH 4.6 (blue)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR