Bromothymol blue ACS

Supplier: Thermo Scientific Chemicals

Synonyms: 3′,3″-Dibromothymolsulphonphthalein, 3′,3″-Dibromothymolsulfonphthalein, 3,3-Bis(3-bromo-4-hydroxy-5-isopropyl-2-methylphenyl)-3H-benzo[c][1,2]oxathiole 1,1-dioxide

32638-06 32638-14
CAAA32638-06EA 53.78 CAD
CAAA32638-06 CAAA32638-14
Bromothymol blue ACS
Bromothymol blue

MDL: MFCD00005872 Beilstein Registry No.: 373934 Soluble in alcohol and aqueous solutions of alkali; sparingly soluble in water


CAS No.: 76-59-5 Molecular Formula: C27H28Br2O5S Formula Weight: 624.39 Physical Form: Crystalline Appearance: Pale brown to red Melting Point: 200-202° Beilstein Reference: 373934 Merck Reference: 14,1445 MDL No.: MFCD00005872

Formula: C₂₇H₂₈Br₂O₅S
Melting Pt: 200…202 °C
Storage Temperature: Ambient
MDL Number: MFCD00005872
CAS Number: 76-59-5
EINECS: 200-971-2
Merck Index: 14,1445

Order Now

Specification Test Results

Clarity of solution Pass test
Visual transition interval pH 6.0 (yellow) to pH 7.6 (blue)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR