Lithium metaborate, anhydrous, tech.
Supplier: Thermo Scientific Chemicals
|
Warning
|
32576-22
32576-36
CAAA32576-22EA
123.71
CAD
CAAA32576-22
CAAA32576-36
Lithium metaborate, anhydrous, tech.
Lithium metaborate
MDL: MFCD00011089
CAS No.: 13453-69-5
Molecular Formula: BLiO2
Formula Weight: 49.75
Physical Form: Powder
Density: 1.397
Sensitivity: Hygroscopic
Appearance: White
Odor: Odorless
Melting Point: 845°
MDL No.: MFCD00011089
Formula:
BLiO₂ Boiling Pt: 1719 °C (1013 hPa) Melting Pt: 845 °C Density: 1.397 Storage Temperature: Ambient |
MDL Number:
MFCD00011089 CAS Number: 13453-69-5 EINECS: 236-631-5 Merck Index: 13,05546 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...