Potassium tetrahydroborate 98%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Potassium tetrahydridoborate, Potassium borohydride

32570-09 32570-22 32570-36 32570-A1
CAAA32570-09EA 42.66 CAD
CAAA32570-09 CAAA32570-22 CAAA32570-36 CAAA32570-A1
Potassium tetrahydroborate 98%
Potassium tetrahydroborate

Source of hydrogen, reducing agent for aldehydes, ketones, acid chlorides; foaming agent for plastics


Purity: 98% CAS No.: 13762-51-1 Molecular Formula: BH4K Formula Weight: 53.94 Physical Form: Powder Density: 1.17 Refractive Index: 1.494 Sensitivity: Moisture Sensitive Appearance: White Odor: Odorless Melting Point: 400° dec. Solubility: Soluble in water (19g/100g ). methanol (3.9g/100g ). 4:1 water:methanol mixture (13g/100g ). Merck Reference: 14,7616 UN No.: 1870 MDL No.: MFCD00011396


Source of hydrogen, reducing agent for aldehydes, ketones, acid chlorides; foaming agent for plastics

Formula: H₄BK
MW: 53.94 g/mol
Melting Pt: >400 °C (dec.)
Density: 1.17
Storage Temperature: Ambient
MDL Number: MFCD00011396
CAS Number: 13762-51-1
EINECS: 237-360-5
UN: 1870
ADR: 4.3,I
Merck Index: 14,7616

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR