Lithium tetrahydroborate ≥95%
Supplier: Thermo Scientific Chemicals
|
Danger
|
Strong reducing agent. Used to reduce compounds containing ketones, aldehydes, ester carbonyls, acid chlorides, lactones, and epoxides
Purity: 95%
CAS No.: 16949-15-8
Molecular Formula: BH4Li
Formula Weight: 21.78
Physical Form: Powder
Density: 0.66
Appearance: White
Odor: Odorless
Melting Point: ca 280° dec.
Solubility: Soluble in ether, THF, and aliphatic amines
Merck Reference: 14,5525
UN No.: 1413
MDL No.: MFCD00011088
Strong reducing agent. Used to reduce compounds containing ketones, aldehydes, ester carbonyls, acid chlorides, lactones, and epoxides
Formula:
H₄BLi Boiling Pt: 66 °C (1013 hPa) Melting Pt: ∼ 280 °C (dec.) Density: 0.66 Storage Temperature: Ambient |
MDL Number:
MFCD00011088 CAS Number: 16949-15-8 EINECS: 241-021-7 UN: 1413 ADR: 4.3,I Merck Index: 14,5525 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...