Glycerine, Ultrapure for spectrophotometry
Supplier: Thermo Scientific Chemicals
|
Warning
|
32450-AE
32450-K2
32450-K7
32450-M1
CAAA32450-AEEA
77.18
CAD
CAAA32450-AE
CAAA32450-K2
CAAA32450-K7
CAAA32450-M1
Glycerine, Ultrapure for spectrophotometry
Glycerine
MDL: MFCD00004722
Beilstein Registry No.: 635685
Common Applications: Useful as a cryoprotectant
CAS No.: 56-81-5
Molecular Formula: HOCH2CH(OH)CH2OH
Formula Weight: 92.09
Flash Point: 160°(320°F)
Physical Form: Viscous liquid
Density: 1.26
Refractive Index: 1.474
Sensitivity: Hygroscopic
Appearance: Colorless
Odor: Odorless
Melting Point: 18°
Boiling Point: 182°/20mm
Beilstein Reference: 635685
Merck Reference: 14,4484
MDL No.: MFCD00004722
Formula:
HOCH₂CH(OH)CH₂OH Boiling Pt: 182 °C (20 mmHg) Melting Pt: 18 °C Density: 1.26 Flash Pt: 160 °C (320 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00004722 CAS Number: 56-81-5 EINECS: 200-289-5 Merck Index: 14,4484 |
|
Specification Test Results
H2O | 0.1% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...