1-Butanol ≥99.0%, Ultrapure for spectrophotometry

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: 1-Butyl alcohol, Butyl Alcohol, 1-Hydroxy butane, Propyl carbinol, n-Butanol, n-Butyl Alcohol

32443-K2 32443-K7 32443-M1
CAAA32443-K2EA 95.12 CAD
CAAA32443-K2 CAAA32443-K7 CAAA32443-M1
1-Butanol ≥99.0%, Ultrapure for spectrophotometry
1-Butanol

MDL: MFCD00002964 Beilstein Registry No.: 969148 Notes: Filtered through 0.2µ filters.


Purity: 99.0+% CAS No.: 71-36-3 Molecular Formula: C4H10O Formula Weight: 74.12 Flash Point: 35°(95°F) Physical Form: Liquid Density: 0.810 Refractive Index: 1.3990 Appearance: Colorless Odor: Alcohol-like pH: 7 Melting Point: -90° Boiling Point: 117-118° Beilstein Reference: 969148 Merck Reference: 14,1540 UN No.: 1120 MDL No.: MFCD00002964

Formula: H₃C(CH₂)₂CH₂OH
MW: 74.12 g/mol
Boiling Pt: 117…118 °C
Melting Pt: –90 °C
Density: 0.810
Flash Pt: 35 °C (95 °F)
Storage Temperature: Ambient
MDL Number: MFCD00002964
CAS Number: 71-36-3
EINECS: 200-751-6
UN: 1120
ADR: 3,III
REACH: 01-2119484630-38
Merck Index: 14,1540
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e9395eeb6150a74222da52460f53cedeccd93442&vid=fd8352a556914c5be7213c576355a88e130d1a7b&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Evaporation residue 5 ppm
H2O 0.1%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR