Chloroform ≥99.5% for spectrophotometry

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Methane, trichloro-, Trichloroform, Methylidyne trichloride, Trichloromethane

32442-K2 32442-K7
CAAA32442-K2EA 107.44 CAD
CAAA32442-K2 CAAA32442-K7
Chloroform ≥99.5% for spectrophotometry
Chloroform

MDL: MFCD00000826 Beilstein Registry No.: 1731042 Notes: Meets ACS Spectrophotometric Requirements. Filtered through 0.2µ filters. Fieser: 1,29 3,140 4,22 5,28 8,92 12,517 15,84


Purity: 99.5+% CAS No.: 67-66-3 Molecular Formula: CHCl3 Formula Weight: 119.38 Physical Form: Liquid Density: 1.492 Refractive Index: 1.4460 Odor: Characteristic Melting Point: -63° Boiling Point: 60.5-61.5° Beilstein Reference: 1731042 Merck Reference: 14,2141 UN No.: 1888 MDL No.: MFCD00000826

Formula: CHCl₃
MW: 119.38 g/mol
Boiling Pt: 61.7
Melting Pt: -63
Density: 1.492
Storage Temperature: Ambient
MDL Number: MFCD00000826
CAS Number: 67-66-3
EINECS: 200-663-8
UN: 1888
ADR: 6.1 III
REACH: 05-2114614906-45
Merck Index: 14,2141
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=9cc00da8cefd4e04c0ea1d7c5801308cd73eeef9&vid=c7d46a2ee73651eae0f3654c272e820106b2f04f&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Evaporation residue 3 ppm
Chlorinated pesticides 5ppt
H2O 0.0004


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR